siRNA Details
Gene Symbol: PELO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC005889.2 | - | 2 | 1529 | 2967612 | |
BC007249.2 | - | 2 | 1529 | 2967002 | ||
BC007650.1 | - | 2 | 1546 | 3608639 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
