siRNA Details
Gene Symbol: CDCP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022842.4 | NP_073753.3 | 6 | 1647 | ||
XM_011534024.1 | XP_011532326.1 | 6 | 1605 | |||
XM_017007070.1 | XP_016862559.1 | 6 | 1633 | |||
GenBank mRNA | AF468010.1 | - | 6 | 1622 | ||
AK023834.1 | - | 4 | 1225 | |||
AK026187.1 | - | 3 | 664 | |||
AK026622.1 | - | 6 | 1622 | |||
AK314441.1 | - | 6 | 1611 | |||
AY026461.1 | - | 6 | 1594 | |||
AY167484.1 | - | 6 | 1604 | |||
AY375452.1 | - | 6 | 1593 | |||
BC069254.1 | - | 6 | 1600 | |||
KU215672.1 | - | 5 | 1425 | |||
KU257676.1 | - | 5 | 1422 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
