siRNA Details
Gene Symbol: LRRC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024512.4 | NP_078788.2 | 4, 5 | 882 | ||
XM_011534110.2 | XP_011532412.1 | 4, 5 | 671 | |||
XM_017007177.1 | XP_016862666.1 | 4, 5 | 882 | |||
GenBank mRNA | AJ308569.2 | - | 4, 5 | 656 | ||
AK057808.1 | - | 4, 5 | 724 | |||
AK223440.1 | - | 4, 5 | 724 | |||
AK314658.1 | - | 4, 5 | 645 | |||
AK315636.1 | - | 4, 5 | 665 | |||
BC029118.1 | - | 4, 5 | 654 | 5183639 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
