siRNA Details
Gene Symbol: UVSSA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317934.1 | NP_001304863.1 | 14 | 2468 | ||
NM_001317935.1 | NP_001304864.1 | 14 | 2456 | |||
NM_020894.3 | NP_065945.2 | 14 | 2569 | |||
XM_017008490.1 | XP_016863979.1 | 14 | 2290 | |||
XM_017008491.1 | XP_016863980.1 | 14 | 2382 | |||
XM_017008492.1 | XP_016863981.1 | 14 | 2380 | |||
XM_017008493.1 | XP_016863982.1 | 14 | 2256 | |||
XM_017008494.1 | XP_016863983.1 | 14 | 2141 | |||
XM_017008495.1 | XP_016863984.1 | 11 | 2283 | |||
XM_017008500.1 | XP_016863989.1 | 8 | 1010 | |||
GenBank mRNA | AB040963.1 | - | 14 | 2569 | ||
AK292661.1 | - | 7 | 1189 | |||
AL110229.1 | - | 1 | 1337 | |||
AL390128.1 | - | 5 | 2723 | |||
BC021930.1 | - | 7 | 1207 | |||
BC110331.1 | - | 14 | 2468 | 5739956 | ||
BC140901.1 | - | 14 | 2306 | 9021769 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
