siRNA Details
Gene Symbol: MELTF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005929.5 | NP_005920.2 | 1, 2 | 169 | ||
NM_033316.3 | NP_201573.1 | 1, 2 | 168 | |||
XM_006713643.3 | XP_006713706.2 | 1, 2 | 168 | |||
XM_011512850.2 | XP_011511152.1 | 1, 2 | 168 | |||
XM_011512851.2 | XP_011511153.1 | 1, 2 | 168 | |||
GenBank mRNA | AA442552.1 | - | 1, 2 | 169 | ||
AU099352.1 | - | 1, 2 | 168 | |||
BC001875.1 | - | 1, 2 | 137 | |||
BC002623.2 | - | 1, 2 | 123 | |||
BC007550.1 | - | 1, 2 | 137 | |||
BC071910.1 | - | 1, 2 | 128 | |||
BC093920.1 | - | 1, 2 | 118 | |||
BC111947.1 | - | 1, 2 | 118 | |||
BC143257.1 | - | 1, 2 | 119 | |||
BT007377.1 | - | 1, 2 | 55 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
