siRNA Details
Gene Symbol: MELTF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005929.5 | NP_005920.2 | 2 | 205 | ||
NM_033316.3 | NP_201573.1 | 2 | 204 | |||
XM_006713643.3 | XP_006713706.2 | 2 | 204 | |||
XM_011512850.2 | XP_011511152.1 | 2 | 204 | |||
XM_011512851.2 | XP_011511153.1 | 2 | 204 | |||
GenBank mRNA | A00127.1 | - | 2 | 151 | ||
AA442552.1 | - | 2 | 205 | |||
AK096182.1 | - | 1 | 1426 | |||
AU099352.1 | - | 2 | 204 | |||
BC001875.1 | - | 2 | 173 | |||
BC002623.2 | - | 2 | 159 | |||
BC007550.1 | - | 2 | 173 | |||
BC032752.1 | - | 2 | 510 | |||
BC071910.1 | - | 2 | 164 | |||
BC093920.1 | - | 2 | 154 | |||
BC111947.1 | - | 2 | 154 | |||
BC143257.1 | - | 2 | 155 | |||
BT007377.1 | - | 2 | 91 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
