siRNA Details
Gene Symbol: SLC9B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100874.2 | NP_001094344.1 | 11 | 1456 | ||
NM_139173.3 | NP_631912.2 | 11 | 1456 | |||
NR_047513.1 | 10 | 1328 | ||||
NR_047515.1 | 9 | 1136 | ||||
XM_006714093.3 | XP_006714156.1 | 12 | 1475 | |||
XM_011531622.1 | XP_011529924.1 | 12 | 1439 | |||
XM_011531623.1 | XP_011529925.1 | 11 | 1443 | |||
GenBank mRNA | AF447585.1 | - | 11 | 1433 | ||
AY461581.1 | - | 11 | 1404 | |||
BC022079.1 | - | 9 | 1052 | |||
BC046636.1 | - | 10 | 1252 | |||
BC110794.1 | - | 11 | 1340 | 5745227 | ||
BC132712.1 | - | 11 | 1383 | 40146734 | ||
BC136966.1 | - | 11 | 1383 | 9020958 | ||
BC144443.1 | - | 8 | 921 | |||
DB526003.1 | - | 1 | 121 | |||
DQ003059.1 | - | 11 | 1436 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
