siRNA Details
Gene Symbol: SLC9B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100874.2 | NP_001094344.1 | 10 | 1253 | ||
NM_139173.3 | NP_631912.2 | 10 | 1253 | |||
NR_047513.1 | 9 | 1125 | ||||
NR_047515.1 | 8 | 933 | ||||
XM_006714093.3 | XP_006714156.1 | 11 | 1272 | |||
XM_011531622.1 | XP_011529924.1 | 11 | 1236 | |||
XM_011531623.1 | XP_011529925.1 | 10 | 1240 | |||
GenBank mRNA | AF447585.1 | - | 10 | 1231 | ||
AY461581.1 | - | 10 | 1201 | |||
BC022079.1 | - | 8 | 849 | |||
BC046636.1 | - | 9 | 1049 | |||
BC110794.1 | - | 10 | 1137 | 5745227 | ||
BC132712.1 | - | 10 | 1180 | 40146734 | ||
BC136966.1 | - | 10 | 1180 | 9020958 | ||
BC144443.1 | - | 7 | 718 | |||
DQ003059.1 | - | 10 | 1233 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
