siRNA Details
Gene Symbol: SPRY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258038.1 | NP_001244967.1 | 3 | 2293 | ||
NM_001258039.1 | NP_001244968.1 | 2 | 2037 | |||
NM_005841.2 | NP_005832.1 | 2 | 2214 | |||
NM_199327.2 | NP_955359.1 | 2 | 2047 | |||
XM_005262686.1 | XP_005262743.1 | 3 | 2954 | |||
XM_017007635.1 | XP_016863124.1 | 2 | 2027 | |||
GenBank mRNA | AF041037.1 | - | 1 | 1279 | ||
AK026960.1 | - | 2 | 2191 | |||
AY590694.1 | - | 1 | 1845 | |||
BC063856.1 | - | 2 | 2176 | 6181674 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
