siRNA Details
Gene Symbol: NAA15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057175.3 | NP_476516.1 | 10 | 1285 | ||
XM_005263236.2 | XP_005263293.1 | 10 | 1340 | |||
GenBank mRNA | AF327722.1 | - | 10 | 1308 | ||
AJ314788.1 | - | 10 | 1237 | |||
AK023387.1 | - | 10 | 1006 | |||
AK023402.1 | - | 10 | 1308 | |||
AK304417.1 | - | 9 | 966 | |||
AK314626.1 | - | 10 | 1340 | |||
AY039242.1 | - | 10 | 1310 | |||
AY112670.1 | - | 10 | 1029 | |||
BC032642.1 | - | 10 | 1310 | |||
BC039818.1 | - | 10 | 1300 | |||
BC093928.1 | - | 10 | 1094 | 7939773 | ||
BC104806.1 | - | 10 | 1094 | 8143809 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
