siRNA Details
Gene Symbol: NAA15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057175.3 | NP_476516.1 | 20 | 4936 | ||
XM_005263236.2 | XP_005263293.1 | 20 | 4988 | |||
GenBank mRNA | AJ314788.1 | - | 20 | 4887 | ||
AK001595.1 | - | 2 | 2387 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
