siRNA Details
Gene Symbol: LMBRD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007527.1 | NP_001007528.1 | 18 | 2854 | ||
XM_011514162.2 | XP_011512464.1 | 18 | 3296 | |||
XM_017010024.1 | XP_016865513.1 | 19 | 2972 | |||
GenBank mRNA | AK091295.1 | - | 18 | 2794 | ||
AL137370.1 | - | 7 | 970 | |||
BC110506.2 | - | 18 | 2793 | 40028737 | ||
BX647340.1 | - | 18 | 2829 | |||
CR749399.1 | - | 18 | 2853 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
