siRNA Details
Gene Symbol: AFAP1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146337.2 | NP_001139809.1 | 18 | 3082 | ||
NM_001323062.1 | NP_001309991.1 | 18 | 3112 | |||
NM_152406.3 | NP_689619.1 | 19 | 3211 | |||
XM_011537558.1 | XP_011535860.1 | 15 | 2761 | |||
XM_017009036.1 | XP_016864525.1 | 19 | 3188 | |||
GenBank mRNA | AK074185.1 | - | 19 | 3154 | ||
AK094067.1 | - | 10 | 2241 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
