siRNA Details
Gene Symbol: AFAP1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146337.2 | NP_001139809.1 | 6 | 593 | ||
NM_001323063.1 | NP_001309992.1 | 6 | 593 | |||
NM_152406.3 | NP_689619.1 | 6 | 593 | |||
XM_011537558.1 | XP_011535860.1 | 2 | 143 | |||
XM_017009036.1 | XP_016864525.1 | 6 | 570 | |||
GenBank mRNA | AK074185.1 | - | 6 | 536 | ||
AK095980.1 | - | 6 | 547 | |||
BC040723.2 | - | 6 | 560 | |||
BC125093.1 | - | 6 | 512 | 40118258 | ||
BC125094.1 | - | 6 | 512 | 40118263 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
