siRNA Details
Gene Symbol: MAP1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324255.1 | NP_001311184.1 | 4 | 1160 | ||
NM_005909.4 | NP_005900.2 | 5 | 1430 | |||
GenBank mRNA | AK096273.1 | - | 6 | 1253 | ||
AK308147.1 | - | 5 | 1221 | |||
BC017240.2 | - | 5 | 1407 | |||
BC025240.1 | - | 5 | 1349 | |||
BC032516.1 | - | 6 | 1273 | |||
BC039822.1 | - | 5 | 1216 | |||
BC046114.1 | - | 5 | 1236 | |||
BC056145.1 | - | 5 | 1210 | |||
BC063669.1 | - | 5 | 1210 | |||
BC073993.1 | - | 5 | 1212 | |||
BC094834.1 | - | 5 | 1207 | |||
BC108733.1 | - | 5 | 1225 | |||
BC139918.1 | - | 5 | 1327 | |||
BC141853.1 | - | 4 | 1013 | |||
BC150196.1 | - | 5 | 1327 | |||
BX647077.1 | - | 5 | 1221 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
