siRNA Details
Gene Symbol: MAP1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324255.1 | NP_001311184.1 | 4 | 5531 | ||
NM_005909.4 | NP_005900.2 | 5 | 5801 | |||
GenBank mRNA | CR933720.1 | - | 1 | 3449 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
