siRNA Details
Gene Symbol: FBXW11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012300.2 | NP_036432.2 | 13 | 3600 | ||
NM_033644.2 | NP_387448.2 | 13 | 3561 | |||
NM_033645.2 | NP_387449.2 | 12 | 3498 | |||
XM_005265855.4 | XP_005265912.1 | 14 | 3650 | |||
XM_005265856.2 | XP_005265913.1 | 15 | 3620 | |||
XM_005265857.2 | XP_005265914.1 | 12 | 3158 | |||
XM_005265858.2 | XP_005265915.1 | 14 | 3662 | |||
XM_017009279.1 | XP_016864768.1 | 13 | 3460 | |||
XM_017009280.1 | XP_016864769.1 | 14 | 3476 | |||
XM_017009281.1 | XP_016864770.1 | 14 | 3544 | |||
GenBank mRNA | AB014596.1 | - | 13 | 3254 | ||
BC026213.1 | - | 13 | 3561 | 4419029 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
