siRNA Details
Gene Symbol: FBXW11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012300.2 | NP_036432.2 | 13 | 2085 | ||
NM_033644.2 | NP_387448.2 | 13 | 2046 | |||
NM_033645.2 | NP_387449.2 | 12 | 1983 | |||
XM_005265855.4 | XP_005265912.1 | 14 | 2135 | |||
XM_005265856.2 | XP_005265913.1 | 15 | 2105 | |||
XM_005265857.2 | XP_005265914.1 | 12 | 1643 | |||
XM_005265858.2 | XP_005265915.1 | 14 | 2147 | |||
XM_017009279.1 | XP_016864768.1 | 13 | 1945 | |||
XM_017009280.1 | XP_016864769.1 | 14 | 1961 | |||
XM_017009281.1 | XP_016864770.1 | 14 | 2029 | |||
GenBank mRNA | AB014596.1 | - | 13 | 1739 | ||
AB033279.1 | - | 12 | 1713 | |||
AB033280.1 | - | 13 | 1831 | |||
AB033281.1 | - | 13 | 1853 | |||
AK294956.1 | - | 13 | 1728 | |||
BC026213.1 | - | 13 | 2046 | 4419029 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
