siRNA Details
Gene Symbol: FBXW11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012300.2 | NP_036432.2 | 6 | 1081 | ||
NM_033644.2 | NP_387448.2 | 6 | 1042 | |||
NM_033645.2 | NP_387449.2 | 5 | 979 | |||
XM_005265855.4 | XP_005265912.1 | 7 | 1131 | |||
XM_005265856.2 | XP_005265913.1 | 8 | 1101 | |||
XM_005265857.2 | XP_005265914.1 | 5 | 639 | |||
XM_005265858.2 | XP_005265915.1 | 7 | 1143 | |||
XM_017009279.1 | XP_016864768.1 | 6 | 941 | |||
XM_017009280.1 | XP_016864769.1 | 7 | 957 | |||
XM_017009281.1 | XP_016864770.1 | 7 | 1025 | |||
GenBank mRNA | AB014596.1 | - | 6 | 735 | ||
AB033279.1 | - | 5 | 709 | |||
AB033280.1 | - | 6 | 827 | |||
AB033281.1 | - | 6 | 849 | |||
AF176022.1 | - | 6 | 734 | |||
AK294956.1 | - | 6 | 724 | |||
AK314999.1 | - | 5 | 608 | |||
BC026213.1 | - | 6 | 1042 | 4419029 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
