siRNA Details
Gene Symbol: C8orf48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007090.2 | NP_001007091.1 | 1 | 438 | ||
GenBank mRNA | AF394908.1 | - | 1 | 291 | ||
AK058131.1 | - | 1 | 438 | |||
BC031245.1 | - | 1 | 386 | 5266320 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
