siRNA Details
Gene Symbol: KIAA1429 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015496.4 | NP_056311.2 | 12 | 2882 | ||
NM_183009.2 | NP_892121.1 | 12 | 2882 | |||
XM_017013306.1 | XP_016868795.1 | 11 | 2662 | |||
XM_017013307.1 | XP_016868796.1 | 11 | 2705 | |||
XM_017013308.1 | XP_016868797.1 | 10 | 2695 | |||
GenBank mRNA | AB037850.1 | - | 12 | 2806 | ||
AF116724.1 | - | 12 | 2806 | |||
AK022906.1 | - | 4 | 653 | |||
AL832487.1 | - | 12 | 2823 | |||
BC003701.1 | - | 5 | 970 | |||
BC069239.1 | - | 5 | 947 | |||
BC112288.1 | - | 12 | 2894 | 8327756 | ||
BC113380.1 | - | 12 | 2882 | 8322432 | ||
BM462567.1 | - | 1 | 42 | |||
CR627454.1 | - | 1 | 426 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
