siRNA Details
Gene Symbol: KIAA1429 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015496.4 | NP_056311.2 | 20 | 4657 | ||
XM_017013306.1 | XP_016868795.1 | 19 | 4437 | |||
XM_017013307.1 | XP_016868796.1 | 19 | 4480 | |||
XM_017013308.1 | XP_016868797.1 | 18 | 4470 | |||
GenBank mRNA | AB037850.1 | - | 20 | 4581 | ||
AK000668.1 | - | 8 | 1363 | |||
AK022906.1 | - | 12 | 2428 | |||
AL117434.1 | - | 7 | 1054 | |||
AL832487.1 | - | 20 | 4598 | |||
BC003701.1 | - | 13 | 2745 | |||
BC069239.1 | - | 13 | 2722 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
