siRNA Details
Gene Symbol: SUGT1P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003667.3 | 4 | 700 | |||
GenBank mRNA | BE264023.1 | - | 4 | 581 | ||
DA804368.1 | - | 4 | 385 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
