siRNA Details
Gene Symbol: SUGT1P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003667.3 | 3, 4 | 654 | |||
GenBank mRNA | BE264023.1 | - | 3, 4 | 535 | ||
DA804368.1 | - | 3, 4 | 339 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
