siRNA Details
Gene Symbol: KCP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017012186.1 | XP_016867675.1 | 40, 41 | 4765 | ||
XM_017012187.1 | XP_016867676.1 | 40, 41 | 4765 | |||
XM_017012188.1 | XP_016867677.1 | 39, 40 | 4458 | |||
XM_017012189.1 | XP_016867678.1 | 39, 40 | 4458 | |||
GenBank mRNA | AK090684.1 | - | 7, 8 | 1089 | ||
AK090715.1 | - | 3, 4 | 1250 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
