siRNA Details
Gene Symbol: HECTD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284274.1 | NP_001271203.1 | 3 | 517 | ||
NM_173497.3 | NP_775768.3 | 3 | 517 | |||
NM_182765.4 | NP_877497.3 | 3 | 517 | |||
NR_104291.1 | 3 | 517 | ||||
XM_017015750.1 | XP_016871239.1 | 3 | 367 | |||
XM_017015751.1 | XP_016871240.1 | 3 | 463 | |||
GenBank mRNA | AK094625.1 | - | 3 | 458 | ||
AK295723.1 | - | 3 | 460 | |||
BC040187.1 | - | 3 | 460 | 4811444 | ||
BX537856.1 | - | 3 | 281 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
