siRNA Details
Gene Symbol: LRRC18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006939.3 | NP_001006940.3 | 1 | 753 | ||
XM_006717867.2 | XP_006717930.1 | 3 | 1230 | |||
XM_011539822.1 | XP_011538124.1 | 4 | 1366 | |||
XM_011539823.1 | XP_011538125.1 | 3 | 1233 | |||
XM_011539826.2 | XP_011538128.1 | 2 | 2520 | |||
XM_011539828.2 | XP_011538130.1 | 2 | 2059 | |||
XM_011539829.2 | XP_011538131.1 | 2 | 2062 | |||
XM_017016276.1 | XP_016871765.1 | 4 | 1369 | |||
XM_017016277.1 | XP_016871766.1 | 2 | 2523 | |||
GenBank mRNA | AY358137.1 | - | 2 | 774 | ||
BC036722.1 | - | 1 | 764 | 5206652 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
