siRNA Details
Gene Symbol: MSMB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002443.3 | NP_002434.1 | 4 | 337 | ||
NM_138634.2 | NP_619540.1 | 3 | 231 | |||
GenBank mRNA | AK312108.1 | - | 4 | 337 | ||
BC005257.1 | - | 4 | 337 | 3950847 | ||
BF680662.1 | - | 3 | 232 | |||
BM856408.1 | - | 4 | 348 | |||
BT006816.1 | - | 3 | 305 | |||
CR457015.1 | - | 3 | 305 | |||
M15885.1 | - | 3 | 316 | |||
S67815.1 | - | 4 | 325 | |||
U22178.1 | - | 2 | 210 | |||
U78976.1 | - | 3 | 329 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
