siRNA Details
Gene Symbol: MSMB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002443.3 | NP_002434.1 | 4 | 471 | ||
NM_138634.2 | NP_619540.1 | 3 | 365 | |||
GenBank mRNA | BC005257.1 | - | 4 | 471 | 3950847 | |
BF680662.1 | - | 3 | 366 | |||
BM856408.1 | - | 4 | 482 | |||
M15885.1 | - | 3 | 450 | |||
S67815.1 | - | 4 | 459 | |||
U22178.1 | - | 2 | 343 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
