siRNA Details
Gene Symbol: RPL36AL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001.4 | NP_000992.1 | 2 | 564 | ||
GenBank mRNA | AB063609.1 | - | 2 | 510 | ||
BC000741.2 | - | 2 | 497 | 3507009 | ||
BC003145.1 | - | 2 | 490 | 2957923 | ||
BC070207.1 | - | 2 | 499 | 4271403 | ||
BC107716.1 | - | 2 | 524 | 6727975 | ||
M15661.1 | - | 2 | 376 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
