siRNA Details
Gene Symbol: TMEM63C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020431.3 | NP_065164.2 | 10 | 882 | ||
GenBank mRNA | AK125159.1 | - | 10 | 814 | ||
BC136613.1 | - | 10 | 764 | 9020603 | ||
BC136614.1 | - | 10 | 761 | 9020604 | ||
BX248756.1 | - | 4 | 258 | |||
BX248759.1 | - | 4 | 229 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
