siRNA Details
Gene Symbol: UCMA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303118.1 | NP_001290047.1 | 4 | 565 | ||
NM_001303119.1 | NP_001290048.1 | 3 | 499 | |||
NM_145314.2 | NP_660357.2 | 5 | 661 | |||
GenBank mRNA | BC018068.1 | - | 5 | 664 | 4799653 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
