siRNA Details
Gene Symbol: TSC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000368.4 | NP_000359.1 | 7 | 835 | ||
NM_001162426.1 | NP_001155898.1 | 7 | 835 | |||
NM_001162427.1 | NP_001155899.1 | 6 | 682 | |||
XM_005272211.1 | XP_005272268.1 | 7 | 880 | |||
XM_006717271.1 | XP_006717334.1 | 7 | 815 | |||
XM_011518979.2 | XP_011517281.1 | 7 | 829 | |||
XM_017015096.1 | XP_016870585.1 | 8 | 1294 | |||
XM_017015097.1 | XP_016870586.1 | 8 | 1265 | |||
XM_017015098.1 | XP_016870587.1 | 7 | 880 | |||
XM_017015099.1 | XP_016870588.1 | 6 | 973 | |||
XM_017015100.1 | XP_016870589.1 | 6 | 944 | |||
XM_017015101.1 | XP_016870590.1 | 6 | 974 | |||
GenBank mRNA | AB190910.1 | - | 7 | 797 | ||
AF013168.1 | - | 7 | 822 | |||
AK297326.1 | - | 6 | 717 | |||
AK299654.1 | - | 5 | 516 | |||
AK303030.1 | - | 6 | 665 | |||
AK308412.1 | - | 7 | 818 | |||
BC047772.2 | - | 7 | 808 | |||
BC070032.1 | - | 7 | 836 | |||
BC108668.1 | - | 7 | 825 | |||
BC121000.2 | - | 7 | 792 | |||
KJ534953.1 | - | 5 | 601 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
