siRNA Details
Gene Symbol: MIR503HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024607.1 | 1, 2 | 87 | |||
GenBank mRNA | BX090522.1 | - | 1, 2 | 87 | ||
CN297454.1 | - | 1, 2 | 64 | |||
KM886853.1 | - | 1, 2 | 36 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
