siRNA Details
Gene Symbol: MIR503HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024607.1 | 2 | 119 | |||
GenBank mRNA | BC007360.2 | - | 3 | 317 | ||
BX090522.1 | - | 2 | 119 | |||
CN297454.1 | - | 2 | 96 | |||
KM886853.1 | - | 2 | 68 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
