siRNA Details
Gene Symbol: OR4K2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005501.1 | NP_001005501.1 | 1 | 907 | ||
GenBank mRNA | BC136941.1 | - | 1 | 943 | 9020933 | |
BC136942.1 | - | 1 | 943 | 9020934 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
