siRNA Details
Gene Symbol: ZNF319 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020807.2 | NP_065858.1 | 2 | 1510 | ||
XM_005256069.3 | XP_005256126.1 | 2 | 1435 | |||
GenBank mRNA | AB037809.1 | - | 2 | 1510 | ||
BC093919.1 | - | 1 | 949 | 7939764 | ||
BC101767.1 | - | 1 | 949 | 8069273 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
