siRNA Details
Gene Symbol: CLDN10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160100.1 | NP_001153572.1 | 2 | 464 | ||
NM_006984.4 | NP_008915.1 | 2 | 353 | |||
NM_182848.3 | NP_878268.1 | 2 | 521 | |||
XM_011521134.2 | XP_011519436.1 | 3 | 446 | |||
XM_017020843.1 | XP_016876332.1 | 3 | 658 | |||
XM_017020844.1 | XP_016876333.1 | 3 | 621 | |||
GenBank mRNA | AK055855.1 | - | 2 | 516 | ||
AK315737.1 | - | 2 | 305 | |||
BC010920.1 | - | 2 | 325 | 4246806 | ||
BU930613.1 | - | 343 | ||||
CR456845.1 | - | 2 | 292 | |||
DA133712.1 | - | 2 | 353 | |||
DA636757.1 | - | 2 | 522 | |||
U89916.1 | - | 2 | 316 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
