siRNA Details
Gene Symbol: CLDN10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160100.1 | NP_001153572.1 | 4 | 648 | ||
NM_006984.4 | NP_008915.1 | 4 | 537 | |||
NM_182848.3 | NP_878268.1 | 4 | 705 | |||
XM_011521134.2 | XP_011519436.1 | 5 | 630 | |||
XM_017020843.1 | XP_016876332.1 | 5 | 842 | |||
XM_017020844.1 | XP_016876333.1 | 5 | 805 | |||
GenBank mRNA | AK055855.1 | - | 4 | 700 | ||
AK315737.1 | - | 4 | 489 | |||
BC010920.1 | - | 4 | 509 | 4246806 | ||
CR456845.1 | - | 4 | 476 | |||
DA133712.1 | - | 4 | 537 | |||
U89916.1 | - | 4 | 500 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
