siRNA Details
Gene Symbol: C4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252204.1 | NP_001239133.1 | 40 | 5260 | ||
NM_007293.2 | NP_009224.2 | 41 | 5398 | |||
GenBank mRNA | AB209989.1 | - | 41 | 5387 | ||
AK293191.1 | - | 16 | 2043 | |||
BC012372.2 | - | 6 | 721 | |||
BC063289.1 | - | 41 | 5431 | 5289068 | ||
BC146673.1 | - | 40 | 5249 | 40148477 | ||
BC151204.1 | - | 41 | 5385 | 40147083 | ||
K02403.1 | - | 41 | 5378 | |||
S81585.1 | - | 6 | 818 |
Gene Symbol: C4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002029.3 | NP_001002029.3 | 41 | 5398 | ||
GenBank mRNA | AK304564.1 | - | 20 | 2513 |
Gene Symbol: C4B_2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242823.2 | NP_001229752.1 | 41 | 5398 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
