siRNA Details
Gene Symbol: TUB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003320.4 | NP_003311.2 | 12 | 1651 | ||
NM_177972.2 | NP_813977.1 | 11 | 1394 | |||
XM_005253109.3 | XP_005253166.1 | 11 | 1386 | |||
XM_011520344.2 | XP_011518646.1 | 11 | 1441 | |||
GenBank mRNA | AK293528.1 | - | 11 | 1322 | ||
BC075031.2 | - | 12 | 1443 | 30915625 | ||
BC075032.2 | - | 12 | 1443 | 30915418 | ||
U54644.1 | - | 11 | 1397 | |||
U82467.1 | - | 12 | 1651 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
