siRNA Details
Gene Symbol: TUB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003320.4 | NP_003311.2 | 13 | 5055 | ||
NM_177972.2 | NP_813977.1 | 12 | 4798 | |||
XM_005253109.3 | XP_005253166.1 | 12 | 4790 | |||
XM_011520344.2 | XP_011518646.1 | 12 | 4845 | |||
GenBank mRNA | BC036400.1 | - | 1 | 2916 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
