siRNA Details
Gene Symbol: ATMIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300728.1 | NP_001287657.1 | 4 | 2308 | ||
NM_015251.2 | NP_056066.2 | 4 | 2368 | |||
GenBank mRNA | AB007891.1 | - | 3 | 2845 | ||
AK025058.1 | - | 1 | 206 | |||
AK290943.1 | - | 4 | 2153 | |||
AK301781.1 | - | 3 | 2108 | |||
BC002701.2 | - | 4 | 2185 | |||
BM693775.1 | - | 1 | 262 | |||
CR749457.1 | - | 4 | 2261 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
