siRNA Details
Gene Symbol: ATMIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300728.1 | NP_001287657.1 | 4 | 2161 | ||
NM_015251.2 | NP_056066.2 | 4 | 2221 | |||
GenBank mRNA | AB007891.1 | - | 3 | 2698 | ||
AK025058.1 | - | 1 | 59 | |||
AK290943.1 | - | 4 | 2006 | |||
AK301781.1 | - | 3 | 1961 | |||
BC002701.2 | - | 4 | 2038 | |||
BM693775.1 | - | 1 | 115 | |||
CR749457.1 | - | 4 | 2114 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
