siRNA Details
Gene Symbol: HSBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001537.3 | NP_001528.1 | 3 | 391 | ||
GenBank mRNA | AF068754.1 | - | 3 | 271 | ||
AK172732.1 | - | 3 | 256 | |||
BC007515.2 | - | 3 | 263 | 3010737 | ||
BX366956.2 | - | 3 | 323 | |||
BX537440.1 | - | 3 | 278 | |||
BX648972.1 | - | 3 | 275 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
