siRNA Details
Gene Symbol: ZNF592 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014630.2 | NP_055445.2 | 4 | 1696 | ||
XM_005254996.3 | XP_005255053.1 | 3 | 1597 | |||
XM_011522246.2 | XP_011520548.1 | 4 | 1766 | |||
XM_011522247.2 | XP_011520549.1 | 3 | 1769 | |||
XM_011522248.2 | XP_011520550.1 | 4 | 1727 | |||
XM_017022734.1 | XP_016878223.1 | 2 | 1477 | |||
GenBank mRNA | BC094688.1 | - | 4 | 1648 | 30397050 | |
BC112232.1 | - | 2 | 1430 | 8327700 | ||
BC112234.1 | - | 2 | 1431 | 8327702 | ||
BC143600.1 | - | 2 | 1432 | |||
D86966.1 | - | 4 | 1930 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
