siRNA Details
Gene Symbol: ZNF592 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014630.2 | NP_055445.2 | 11 | 3976 | ||
XM_005254996.3 | XP_005255053.1 | 10 | 3877 | |||
XM_011522246.2 | XP_011520548.1 | 11 | 4046 | |||
XM_011522247.2 | XP_011520549.1 | 10 | 4049 | |||
XM_011522248.2 | XP_011520550.1 | 12 | 4042 | |||
XM_017022734.1 | XP_016878223.1 | 9 | 3757 | |||
GenBank mRNA | BC094688.1 | - | 11 | 3928 | 30397050 | |
BC112232.1 | - | 9 | 3710 | 8327700 | ||
BC112234.1 | - | 9 | 3711 | 8327702 | ||
BC143600.1 | - | 9 | 3701 | |||
D86966.1 | - | 11 | 4210 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
