siRNA Details
Gene Symbol: PITPNC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012417.3 | NP_036549.2 | 6, 7 | 1344 | ||
NM_181671.2 | NP_858057.1 | 6, 7 | 1344 | |||
XM_005257216.2 | XP_005257273.1 | 6, 7 | 844 | |||
XM_017024442.1 | XP_016879931.1 | 6, 7 | 455 | |||
XM_017024443.1 | XP_016879932.1 | 6, 7 | 455 | |||
XM_017024444.1 | XP_016879933.1 | 6, 7 | 846 | |||
XM_017024445.1 | XP_016879934.1 | 6, 7 | 468 | |||
GenBank mRNA | AF171102.1 | - | 6, 7 | 470 | ||
AK290838.1 | - | 6, 7 | 1166 | |||
AK307138.1 | - | 3, 4 | 608 | |||
BC007905.2 | - | 6, 7 | 817 | 4299595 | ||
BC067095.1 | - | 6, 7 | 1065 | 6088874 | ||
BE906407.1 | - | 4, 5 | 250 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
