siRNA Details
Gene Symbol: PITPNC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012417.3 | NP_036549.2 | 8 | 1546 | ||
NM_181671.2 | NP_858057.1 | 8 | 1546 | |||
XM_005257216.2 | XP_005257273.1 | 8 | 1046 | |||
XM_017024442.1 | XP_016879931.1 | 8 | 657 | |||
XM_017024443.1 | XP_016879932.1 | 8 | 657 | |||
XM_017024444.1 | XP_016879933.1 | 8 | 1048 | |||
XM_017024445.1 | XP_016879934.1 | 8 | 670 | |||
GenBank mRNA | AF171102.1 | - | 8 | 672 | ||
AK290838.1 | - | 8 | 1368 | |||
AK307138.1 | - | 5 | 810 | |||
BC007905.2 | - | 8 | 1019 | 4299595 | ||
BC067095.1 | - | 8 | 1267 | 6088874 | ||
BE906407.1 | - | 6 | 452 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
