siRNA Details
Gene Symbol: PITPNC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012417.3 | NP_036549.2 | 7 | 1416 | ||
NM_181671.2 | NP_858057.1 | 7 | 1416 | |||
XM_005257216.2 | XP_005257273.1 | 7 | 916 | |||
XM_017024442.1 | XP_016879931.1 | 7 | 527 | |||
XM_017024443.1 | XP_016879932.1 | 7 | 527 | |||
XM_017024444.1 | XP_016879933.1 | 7 | 918 | |||
XM_017024445.1 | XP_016879934.1 | 7 | 540 | |||
GenBank mRNA | AF171102.1 | - | 7 | 542 | ||
AK290838.1 | - | 7 | 1238 | |||
AK307138.1 | - | 4 | 680 | |||
BC007905.2 | - | 7 | 889 | 4299595 | ||
BC067095.1 | - | 7 | 1137 | 6088874 | ||
BE906407.1 | - | 5 | 322 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
